Skip to Content
MilliporeSigma
All Photos(1)

Documents

PSN1

94060601, human pancreas, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PSN1, 94060601

biological source

human pancreas

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human pancreatic adenocarcinoma

Cell Line Description

The human pancreatic cancer cell line PSN1 was derived from pancreatic adenocarcinoma tissue. After transplantation into nude mice the PSN1 cell line was established from the xenograft. PSN1 cells are unique in that amplification of both c-myc (50-fold) and activated c-Ki-ras (3-6 fold) with a point mutation from GGT to CGT at codon 12 are present in the same degree as in the original tumour and contain increased amounts of c-myc and c-Ki-ras transcripts. Loss of one of the two p53 alleles and a point mutation from AAG to CAG at codon 132 in the remaining allele was also observed in PSN1.

Application

PSN1 has been used to study synthetic acetylhomoagmatine in vitro. It has also been used to determine the cytotoxicity of irinotecan and topotecan.
PSN1 is an example of a cancer cell having multiple genetic changes including those of two oncogenes (c-Ki-ras and c-myc) and a tumour suppresser gene (p53).

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:8 i.e. seeding at 1-3 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service