Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CTLL-2

93042610, mouse blood, Lymphoblast

Synonym(s):

CTLL-2 cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CTLL-2, 93042610

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C57b1/6 T cell

Cell Line Description

CTLL is a subclone of T cells derived from a C57bl/6 mouse. The cells require IL-2 for growth and are used to assay for its presence in conditioned media.

Application

CTLL-2 has been used in CTLL-2 cell proliferation assay to determine the presence of T-cell cytokines.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10-20 IU/ml Human Recombinant IL-2 + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml; 5% CO2; 37°C. Thaw ampoule and perform a cell count, spin out the DMSO and resuspend in an appropriate volume of media to give a cell concentration of 4-5 x 105 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service