Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ND-C

92090913, mouse nerve, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

ND-C, 92090913

biological source

mouse nerve

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat neurone hybrid

Cell Line Description

ND-C hybrid cells were derived after fusion of primary neonatal-rat dorsal-root-gauglion (DRG) neurons with N18Tg2, a mouse neuroblastoma (C1300) derived azaguanine resistant line. HAT sensitive hybrids were selected and cloned by limiting dilution. The cells express rat Thy 1.1 marker and the sensory neuropeptide substance P. The cell line has been mycoplasma eradicated at ECACC.

Application

Study of neuronal differentiation, cell adhesion and expression of neuronal proteins.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3 x 10,000 cells/cm2; aspirate or gently tap cells loose, use of trypsin not necessary; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service