Skip to Content
MilliporeSigma
All Photos(1)

Documents

SH-SY5Y Cell Line human

94030304, human nerve, Neuroblast

Synonym(s):

SHSY5Y Cells, SK-SH-SY5Y Cells, SY5Y Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SH-SY5Y Cell Line human, 94030304, Neuroblast from neural tissue.

biological source

human nerve

growth mode

Adherent

karyotype

Not specified

morphology

Neuroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human neuroblastoma

Cell Line Description

SH-SY5Y is a thrice-cloned sub-line of bone marrow biopsy-derived line SK-N-SH (Sigma catalogue no. 86012802). SH-SY-5Y has dopamine-β-hydroxylase activity and can convert glutamate to the neurotransmitter GABA. Will form tumours in nude mice in approximately 3-4 weeks. The loss of neuronal characteristics has been described with increasing passage numbers. Therefore it is recommended to verify specific characteristics such as noradrenalin uptake or neuronal markers routinely.

Application

SH-SY5Y cell line human is used for neurotransmitter studies
SH-SY5Y cell line human has been used to:
  • study the iron-mediated toxicity of amyloid β peptide
  • check the significance of anionic luminescent conjugated polythiophene (LCP) polythiophene acetic acid (PTAA) in vital staining of cells
  • study the effect of haloperidol on sigma-1 receptors in guinea pig brain and human neuroblastoma cells

Other Notes

  • DNA is stored at -70 °C
  • RNA is stored at -70 °C
  • Cell line is stored at -196 °C
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11
D16S539: 8,13
D5S818: 12
D7S820: 7,10
THO1: 7,10
TPOX: 8,11
vWA: 14,18

Culture Medium

Ham′s F12:EMEM (EBSS) (1:1) + 2mM Glutamine + 1% Non Essential Amino AcidsNEAA + 15% Foetal Bovine Serum FBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:100 i.e. seeding at 1x1,000-1x10,000 cells/cm2; using 0.25% Trypsin or Trypsin/EDTA, 5% CO2; 37°C. Cells may reattach slowly and may remain in suspension for several days. The loss of neuronal characteristics has been described with increasing passage numbers. Therefore it is recommended to verify specific characteristics such as noradrenalin uptake or neuronal markers routinely.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service