Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G-361

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 88030401, human skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human skin

packaging

tube of 5 μg 88030401-DNA-5UG
pkg of vial of cells 88030401-1VL

growth mode

Adherent

karyotype

Triploid, modal n.o 69

morphology

Epithelial

products

Melanin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian malignant melanoma

Cell Line Description

Established from a malignant melanoma of a 31 year old male Caucasian. The cells produce melanin for up to 50 population doublings. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Application

Hormone studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11
D16S539: 15
D5S818: 11
D7S820: 8,11
THO1: 7,9
TPOX: 8,10
vWA: 15,17

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Cells are slow in attaching.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service