Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCC257

Sigma-Aldrich

OVCAR-3 Human Ovarian Cancer Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.28

description

Cell line type: Cancer Cells

technique(s)

cell culture | mammalian: suitable

General description

Ovarian carcinoma is a prevalent worldwide disease, with over 200,000 new cases diagnosed per year (1). The first-line chemotherapy for newly-diagnosed ovarian carcinoma is cisplatin, but despite high initial efficacy, the most common types of ovarian carcinomas frequently relapse into platinum-resistant forms and many demonstrate multi-drug resistance (1). The availability of cellular models that recapitulate both resistant and susceptible forms of the disease are essential to understanding drug resistance and advancing new options for treatment.

OVCAR-3 is a high-grade serous ovarian adenocarcinoma cell line established from a patient refractory to cisplatin, however the cell line remains sensitive to a variety of chemotherapeutic drugs (2). OVCAR-3 cells demonstrate increased glutathione content (3) and express the Wilms Tumor 1 protein (WT1), a marker of advanced serous ovarian carcinoma (4). OVCAR-3 cells are characterized by migration/invasion ability and are tumorigenic in nude mice (5). The OVCAR-3 cell line is well-established and one of the most highly-cited model systems for ovarian carcinoma.

Source:
OVCAR-3 was established from ascitic fluid from a 60-year-old patient with ovarian cancer refractory to cisplatin treatment (2).

References:
1. Nat Rev Disease Primers 2016; 2:16061.
2. Cancer Res 1997; 57:850—856.
3. Proc Natl Acad Sci USA 1992; 89:3070-3074.
4. BMC Cancer 2006; 6:90.
5. Gynecol Oncol 2015;138(2):372-377.

Application

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

  • Each vial contains >=1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for inter-species contamination from rat, mouse, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are negative for mycoplasma contamination.
  • Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service