Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IMR 32

86041809, human nerve, Fibroblast/neuroblast

Synonym(s):

IMR32 Cells, Institute for Medical Research-32 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

IMR 32, 86041809

biological source

human nerve

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast/neuroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian neuroblastoma

Cell Line Description

Established from an abdominal mass occurring in a 13 month old Caucasian male. The tumour was diagnosed as a neuroblastoma with two rare areas of orgonoid differentiation. The culture is a mixture of two morphologically distinct cell types, a small neuroblast-like cell and the other a large hyaline fibroblast. IMR 32 is partially resistant to polioviruses type 3 and highly resistant to ECHO-II. It is susceptible to Coxsackie B3.

Application

IMR 32 has been used to study the transmigration of cells through a human in vitro blood-cerebrospinal fluid barrier (BCSFB) model.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. The cells have a low plating efficiency and cell attachment can take up to 2-4 days after resuscitation. Cells easily detach. May n

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service