Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WiDr

85111501, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human colon
growth mode:
Adherent
karyotype:
2n = 46, hypertriploid
morphology:
Epithelial
products:
Carcinoembryonic antigen (CEA)
receptors:
Not specified
relevant disease(s):
cancer

Product Name

WiDr, 85111501

biological source

human colon

growth mode

Adherent

karyotype

2n = 46, hypertriploid

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

Although originally described as a colon adenocarcinoma line established from a 78 year old female, DNA fingerprinting has shown this line to be a derivative of HT-29. HT29 was isolated from a primary colon adenocarcinoma in a 44 year old Caucasian female. The cells produce carcinoembryonic antigen and are tumorigenic in xenogeneic animals. They have been used in studies of anti-tumour agents.

Application

Carcinogenesis and tumourigenicity studies and anti-tumour testing

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 11,12
D16S539: 11,12
D5S818: 11,12
D7S820: 10
THO1: 6,9
TPOX: 8,9
vWA: 17,19 STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 11,1

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service