Skip to Content
MilliporeSigma
All Photos(1)

Documents

COLO 201

87091201, human colon. Clumps of spherical cells in suspension, cuboidal cells in monolayer.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COLO 201, 87091201

biological source

human colon

growth mode

Adherent

karyotype

Triploid modal no. 71-78

morphology

Clumps of spherical cells in suspension, cuboidal cells in the monolayer

products

Carcinoembryonic antigen (CEA) less than produced by COLO 205

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

COLO 201, COLO 205 and COLO 206 (ECACC Catalogue 93052620) were all established from the ascites fluid obtained from a 70-year-old male patient with adenocarcinoma of the colon (Dukes Classification Grade D). At time of isolation, COLO 201 displayed cuboidal morphology but with increasing passage the depositor reported that COLO 201 tends to display predominantly rounded cells in clumps in suspension with cuboidal cells in the monolayer. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 10
D16S539: 12,13
D5S818: 10,13
D7S820: 9,10
THO1: 8,9
TPOX: 11
vWA: 15

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain suspension cells at 3-9x105 cells/cm3. Split sub-confluent cultures (70-80%) of attached cells 1:3 to 1:6 i.e. seeding at 2-3x104 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Diverse colorectal cancer cell lines with relevant mutations in genes KRAS, APC, TP53, and PTEN for drug delivery and metastasis studies.

Diverse colorectal cancer cell lines with relevant mutations in genes KRAS, APC, TP53, and PTEN for drug delivery and metastasis studies.

Diverse colorectal cancer cell lines with relevant mutations in genes KRAS, APC, TP53, and PTEN for drug delivery and metastasis studies.

Diverse colorectal cancer cell lines with relevant mutations in genes KRAS, APC, TP53, and PTEN for drug delivery and metastasis studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service