Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14650C

SAFC

EX-CELL® NS0 Serum-Free Medium for NS0 Cells

Animal-component free, Protein-free, Chemically defined, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

glucose: 5.6 g/L
L-glutamine: no
HEPES: no
NaHCO3: 2.14 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL NS0 is an animal-component free, protein-free, chemically defined, serum-free medium developed for the long-term growth of NS0-related cells in suspension culture. The NS0 cells are clonal derivatives of the parent NS1 cell line and are capable of growth in suspension culture. NS0 hybridoma suspension cultures can be subcultured direclty into EX-CELL NS0 from serum-supplemented or serum-free media. Suspension cultures in EX-CELL NS0 have been carried for more that 50 passages with no loss of growth or viability.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mladen Korbelik et al.
International immunopharmacology, 75, 105764-105764 (2019-07-29)
It is becoming apparent that to obtain robust and prolonged antitumor responses in cancer immunotherapy, appropriate adjunct agents promoting both tumor antigen delivery and immune rejection enhancement are critically required. The semisynthetic biopolymer N-dihydrogalactochitosan (GC) is emerging as a promising

Articles

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service