Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAJI′A′

85022102, human lymph, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human lymph
growth mode:
Suspension
karyotype:
Modal no. 47-48
morphology:
Fibroblast-like
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

RAJI′A′, 85022102

biological source

human lymph

growth mode

Suspension

karyotype

Modal no. 47-48

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Burkitt′s lymphoma

Cell Line Description

Clone of RAJI selected for adherence to glass or plastic. Fibroblast like cell line. RAJI ′A′ was developed from the adherent sub-population of RAJI cells. Karyotype analysis demonstrated that RAJI ′A′ contains typical human chromosomes and the modal total chromosome number (47-48) is the same as that of RAJI. RAJI ′A′ is also identical to RAJI with respect to isoenzyme composition, susceptibility to EBV infection and inducibility of latent EBV. Ethnicity: Black.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 13
D16S539: 8,11
D5S818: 10,13
D7S820: 10
THO1: 6,7
TPOX: 8,13
vWA: 17,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service