Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-337-M

Sigma-Aldrich

MAPKAP Kinase 2 Protein, active, 10 µg

Active, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in E. coli Human MAPKAP Kinase
expressed in E. coli

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 70.2 kDa

purified by

(GSH-agarose & Q-Sepharose FF)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

accession no.

 (FUNCTION: SwissProt: P49137 # Its physiological substrate seems to be the small heat shock protein (HSP27/HSP25). In vitro can phosphorylate glycogen synthase at ′Ser-7′ and tyrosine hydroxylase (on ′Ser-19′ and ′Ser-40′). This kinase phosphorylates Ser in the peptide sequence, Hyd-X-R-X(2)-S, where Hyd is a large hydrophobic residue (By similarity). Mediates both ERK and p38 MAPK/MAPK14 dependent neutrophil responses. Participates in TNF alpha-stimulated exocytosis of secretory vesicles in neutrophils. Plays a role in phagocytosis-induced respiratory burst activity. )
NM_004759.3

NCBI accession no.

UniProt accession no.

Gene Information

human ... MAPKAPK2(9261)

General description

Product Source: Human MAPKAP Kinase 2 expressed in E. coli.
Recombinant human MAPKAP Kinase 2, residues 46-400, A399G, containing an N-terminal GST tag.

MAPKAP Kinase 2 has been implicted as a cell cycle checkpoint kinase, joining the ranks of Chk1 and Chk2 as critical regulators of the DNA damage response in mammalian cells in recent studies (Manke, et al, 2005, and Abraham, 2005). This research indicates that MAPKAP Kinase 2, a serine/threonine kinase activated by p38, phosphorylates CDC25 B/C, generating a binding site for 14-3-3 proteins. The checkpoint kinases Chk1 and Chk2 are targets for drug discovery efforts designed to overcome cell cycle arrest due to DNA damage induced by chemotherapeutic agents. Overriding the checkpoint kinases in cancer cells exposed to DNA damaging agents leads to catastrophic failure of cell division and apoptosis.
The new role of MAPKAP Kinase 2 in complementing the function of Chk1 and Chk2 suggests a new target for drug discovery efforts.
This gene encodes a member of the Ser/Thr protein kinase family. This kinase is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, this kinase is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for this gene.

Biochem/physiol Actions

Protein Target: MAPKAP-K2
Target Sub-Family: CAMK

Packaging

Also available in 10 μg size and in bulk
Also available in 250μg size as catalog # 14-337M - Call for pricing and availability.

Quality

Routinely evaluated by phosphorylation of MAPKAP Kinase 2 Substrate Peptide

Target description

MAPKAP-K2

Physical form

GSH-agarose & Q-Sepharose FF

Storage and Stability

1 year at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Xu et al.
The Biochemical journal, 355(Pt 2), 437-447 (2001-04-04)
When fibroblasts are cultured in contracting collagen matrices, matrix metalloproteinase-1 (MMP-1, collagenase-1) is induced. In the present study we demonstrate that p38alpha mitogen-activated protein kinase (p38alpha MAPK) plays a bi-directional role in the MMP-1 response to contracting floating collagen lattices
Madhavi J Rane et al.
The Journal of biological chemistry, 278(30), 27828-27835 (2003-05-13)
Activation of the serine-threonine kinase Akt by cytokines, chemokines, and bacterial products delays constitutive neutrophil apoptosis, resulting in a prolonged inflammatory response. We showed previously that Akt exists in a signaling complex with p38 MAPK, MAPK-activated protein kinase-2 (MAPKAPK-2), and
Y L Woods et al.
The Biochemical journal, 355(Pt 3), 597-607 (2001-04-20)
Forkhead in rhabdomyosarcoma (FKHR) is a transcription factor that has been implicated in the control of gene expression by insulin, as well as the regulation of apoptosis by survival factors. These signals trigger the protein kinase B (PKB)-catalysed phosphorylation of
M Suomalainen et al.
The EMBO journal, 20(6), 1310-1319 (2001-03-17)
Nuclear targeting of adenovirus is mediated by the microtubule-dependent, minus-end-directed motor complex dynein/dynactin, in competition with plus- end-directed motility. We demonstrate that adenovirus transiently activates two distinct signaling pathways to enhance nuclear targeting. The first pathway activates integrins and cAMP-dependent
D Stokoe et al.
The EMBO journal, 11(11), 3985-3994 (1992-11-01)
A novel protein kinase, which was only active when phosphorylated by the mitogen-activated protein kinase (MAP kinase), has been purified 85,000-fold to homogeneity from rabbit skeletal muscle. This MAP kinase activated protein kinase, termed MAPKAP kinase-2, was distinguished from S6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service