Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3B9

00020913, Reactivity: Anti-Acetylated α-Tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

Mouse spleen

Myeloma

Sp2-0/Ag

Reactivity

Anti-Acetylated α-Tubulin

technique(s)

cell culture | mammalian: suitable

isotype

IgG12b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

C3B9 was developed by immunizing mice with extracted T. brucei brucei cytoskeleton and fusing the mouse spleen cells with the myeloma Sp2-0/Ag.

Immunogen

T. brucei brucei cytoskeleton (detergent extracted)

Application

Detection of acetylated α tubulin.

Subculture Routine

Maintain cultures between 1-9 x 100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service