Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IMR-90 Cell Line human

85020204, human lung (fetal), Fibroblast

Synonym(s):

I90 Cells, IMR90 Cells, Institute for Medical Research-90 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

IMR-90 Cell Line human, 85020204

biological source

human lung (fetal)

description

Human Caucasian fetal lung fibroblast

growth mode

Adherent

karyotype

2n = 46, diploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian foetal lung fibroblast

Cell Line Description

Derived from lung tissue of a 16 week old female Caucasian foetus. The cells have a virus susceptibility similar to WI-38 (Sigma catalogue no. 90020107) and MRC-5 cells (Sigma catalogue no. 84101801). The cells have a finite lifespan. Available as a growing order only.

Application

IMR-90 human cell line has been used to study the cytotoxicity of hydroxyapatite/gold/arginine (HAp/Au/arginine) nanoparticles.
Virus studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 11,13
D16S539: 10,13
D5S818: 12,13
D7S820: 9,12
THO1: 8,9.3
TPOX: 8,9
vWA: 16,19

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service