Skip to Content
MilliporeSigma
All Photos(1)

Documents

LCL721 human cell line

IHW Number: 9249, 09071301

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

ethnicity

Not specified

gender

female

growth mode

suspension

karyotype

Not specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Consanguineous: No
Homozygous: No

Cell Line Description

An Epstein-Barr virus (EBV) transformed lymphoblastoid cell line with Human Leukocyte Antigen (HLA)-typing data available.

Culture Medium

RPMI 1640 (R0883) + 2mM L-Glutamine (G7513) + 10% FBS (F2442). To help establish a culture the concentration of FBS can be increased to 20%.

Subculture Routine

When resuscitating from frozen, cells should be seeded at 0.5 x 106 cells/ml in the culture medium. The culture should establish log phase growth after 3-5 days. When growing well dilute every 1-3 days to 3 x 105 cells/ml. Maintain cultures between 3-9 x 105 cells/ml for maximum growth; 5% CO2; 37°C. Cells form floating aggregates of varying size.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service