Skip to Content
MilliporeSigma
All Photos(1)

Documents

h1RPE7

9061602, human eye, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

h1RPE7, 09061602, from human eye

biological source

human eye

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal

products

Cytokeratins 5 and 8, junctional protein zonula occludens (ZO-1)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Origin

An immortalized retinal pigment epithelial cell line

Cell Line Description

The h1RPE7 cell line was derived from primary human retinal pigment epithelial cells obtained from a 50-year-old female Caucasian donor. The primary culture was transfected with a construct encoding SV40 large T antigen that was deleted for the small t antigen along with puromycin resistance gene.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 10,13
D13S317: 12
D16S539: 12
D5S818: 9,13
D7S820: 9,10
THO1: 7,9.3
TPOX: 8
vWA: 16,19

Culture Medium

Hams F10 + 20% Heat-inactivated FCS + 2 mM Glutamine + selection antibiotic 1μg/ml puromycin.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 , i.e., seeding at 3 x 10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Population doubling approx 24hrs.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service