Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7

7021402, mouse lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

T7, 07021402

biological source

mouse lung

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Type-II lung epithelial cell

Cell Line Description

Fully differentiated Type-II alveolar epithelial cell line derived from transgenic H-2Kb-tsA58 mice capable of being passaged as an immortalised cloned cell line in culture. Cells will proliferate at 33 - 35°C in the presence of gamma-interferon, at 39°C in the absence of gamma-interferon cells will terminally differentiate. Supplied as frozen ampoules only.

Culture Medium

DMEM + 2mM Glutamine + 4% Foetal Bovine Serum (FBS) + 8 additional supplements - please see ′Additional Literature Report′ field below for full details. ECACC can supply an information sheet on the preparation of the supplements on request - contact ECAC

Subculture Routine

Split confluent cultures 1:5 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA;10% CO2; 33°C. Doubling time 48h

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service