Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KARPAS 422

06101702, human lymph (lymphoma). Round polygonal singlets or small clusters in suspension

Synonym(s):

KARPAS, 422

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

KARPAS 422, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., from human lymph(lymphoma), 06101702

biological source

human lymph (lymphoma)

packaging

tube of 5 μg 06101702-DNA-5UG
pkg of vial of cells 06101702-1VL

growth mode

Suspension

karyotype

Hyperdiploid with 10% polyploidy

morphology

Round polygonal cells growing singly or in small clusters in suspension

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human B cell non-Hodgkin lymphoma

Cell Line Description

Established from a pleural effusion of 73 year-old woman diagnosed with B-cell non-Hodgkin lymphoma (intra-abdominal, diffuse large cell lymphoma, refractory, terminal). Carries t(14;18) IGH-BCL2 fusion gene (break point in major breakpoint region, MBR).

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 20% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 0.5 - 2 x 1000,000 cells/ml; 5% CO2; 37 °C. Doubling time approximately 60-90 hours. Split saturated culture 1:2 every 2-4 days. Round polygonal cells growing singly or in small clusters in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service