Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS4870

Corning® Costar® reagent reservoirs

white polystyrene, capacity 50 mL

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.13

material

U-bottom bottom
white polystyrene

sterility

sterile

packaging

pkg of 200 ea
pkg of 5 ea

manufacturer/tradename

Corning 4870

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Costar® reagent disposable reservoirs are used to repetitively fill multichannel pipettors. These reserviors are non-pyrogenic and free of DNase/RNase contamination.

Application

Corning® Costar reagent reservoirs are suitable for use with multi-channel pipettors to transfer samples, buffers, or reagents into enzyme-linked immunosorbent assay(ELISA) microplates.

Features and Benefits

Costar®reagent reservoirs are manufactured from modified polystyrene, are sterile, anddisposable.

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service