Skip to Content
MilliporeSigma

C6942

Nalgene® round carboy with spigot

nominal capacity 25 L (6.5 gal), low-density polyethylene

Synonym(s):

carboys, nalgene bottles, nalgene containers, Nalgene 2318-0065, laboratory container, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41121820
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

low-density polyethylene , polypropylene spigot and screw closure

packaging

pack of 1 ea

manufacturer/tradename

Nalgene 2318-0065

H × D

23 1/2 in. (597 mm) × 11 3/8 in. (287 mm)

nominal capacity

25 L (6.5 gal)

General description

Designed for easy dispensing. Graduated in 1 gallon and 5 litre increments. Spigot boss molded in one piece with carboy. Spigot closure included. 83B (83 mm) closure except where noted.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nalgene® round carboy with spigot




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library