Key Documents
Z675563
Polypropylene carboy
size 20 L
About This Item
UNSPSC Code:
41121820
NACRES:
NB.35
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F10794-0050
size
20 L
General description
All polypropylene construction is autoclavable 121°C (250°F) and chemical resistant. When autoclaving, do not install cap.
  • 83 mm cap seals with a silicone gasket
  • One large handle for transport or pouring
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)