Skip to Content
MilliporeSigma

C5567

Nalgene® round carboy, with handles

nominal capacity 50 L (13 gal), polypropylene

Synonym(s):

carboys, nalgene carboys, Nalgene 2250-0130, laboratory container, receptical, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41121820
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene , white polypropylene screw closure

packaging

pack of 1 ea

manufacturer/tradename

Nalgene 2250-0130

H × D

26 1/2 in. (673 mm) × 14 7/8 in. (376 mm)

nominal capacity

50 L (13 gal)

General description

Graduated in 1 gal and 5 L increments. 83B (83 mm) closure.
Screw closure with thermoplastic elastomer (TPE) gasket. Autoclavable (only when empty). Not recommended for use with hazardous materials.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library