Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C211850

Sigma-Aldrich

DMT-2′O-MOE-rMeC(bz) Phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H60N5O10P
CAS Number:
Molecular Weight:
922.01
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51

Assay

≥98.0% (reversed phase HPLC)

Quality Level

form

powder

λ

conforms (UV/VIS Identity)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-MOE-rMeC(bz) Phosphoramidite is a phosphoramidite that belongs to the family of amides of trivalent phosphorus acid H3PO3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service