Skip to Content
MilliporeSigma

Skip To

A702000

DMT-rA(tac)-CPG

pore size 1000 Å, 20-30 μmol/g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.51
UNSPSC Code:
12352200
Assay:
≥98.0% (Nucleoside Purity, reversed phase HPLC)
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

1000 Å

capacity

≥98 % coupling capacity (Coupling Efficiency)

storage temp.

−20°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T401000G211850A211850
Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

300

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (reversed phase HPLC)

assay

≥98.0% (reversed phase HPLC)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

pore size

1000 Å

pore size

1000 Å

pore size

-

pore size

-

capacity

≥98 % coupling capacity (Coupling Efficiency)

capacity

≥99 % (Coupling Efficiency)

capacity

-

capacity

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters