Key Documents
B5910
Sigma-Aldrich
Nalgene® bottles, style 2007
capacity 500 mL
Synonym(s):
HDPE bottles, nalgene bottle, nalgene bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
high-density polyethylene bottle
polypropylene cap
polypropylene screw closure
rectangular translucent bottle
Joint
Joints threaded neck (48-415)
description
Not autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene 2007-0016
bottle L × H × W
94 mm × 117 mm × 58.7 mm
capacity
500 mL
General description
Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. and have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)