Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV50600

Sigma-Aldrich

Anti-SENP6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ11355, Anti-FLJ11887, Anti-KIAA0389, Anti-KIAA0797, Anti-SSP1, Anti-SUMO1/Sentrin specific peptidaSe 6, Anti-SUSP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

125 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SENP6(26054)

General description

SENP6 is a SUMO-deconjugating enzyme that negatively modulates TLR-mediated inflammatory signaling. Studies have reported that SENP6 also regulates PML nuclear bodies and is required for inner kinetochore assembly.
Rabbit Anti-SENP6 antibody recognizes human SENP6.

Immunogen

Synthetic peptide directed towards the C terminal region of human SENP6

Application

Rabbit Anti-SENP6 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

SENP6 is a UBL-specific protease that deconjugates SUMO1, SUMO2 and SUMO3 from targeted proteins. It does not seem to be involved in the processing of full-length SUMO proteins to their mature forms. SENP6 deconjugates SUMO1 from RXRA, leading to transcriptional activation. It may act preferentially on substrates containing 3 or more SUMO2 or SUMO3 moieties.Ubiquitin-like molecules (UBLs), such as SUMO1 (UBL1; MIM 601912), are structurally related to ubiquitin (MIM 191339) and can be ligated to target proteins in a similar manner as ubiquitin. However, covalent attachment of UBLs does not result in degradation of the modified proteins. SUMO1 modification is implicated in the targeting of RANGAP1 (MIM 602362) to the nuclear pore complex, as well as in stabilization of I-kappa-B-alpha (NFKBIA; MIM 164008) from degradation by the 26S proteasome. Like ubiquitin, UBLs are synthesized as precursor proteins, with 1 or more amino acids following the C-terminal glycine-glycine residues of the mature UBL protein. Thus, the tail sequences of the UBL precursors need to be removed by UBL-specific proteases, such as SENP6, prior to their conjugation to target proteins (Kim et al., 2000 [PubMed 10799485]).[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: MNLANWFPPPRMRTKREEIRNIILKLQEDQSKEKRKHKDTYSTEAPLGEG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing Liu et al.
PLoS pathogens, 9(6), e1003480-e1003480 (2013-07-05)
The signaling of Toll-like receptors (TLRs) induces host defense against microbial invasion. Protein posttranslational modifications dynamically shape the strength and duration of the signaling pathways. It is intriguing to explore whether de-SUMOylation could modulate the TLR signaling. Here we identified
Debaditya Mukhopadhyay et al.
The Journal of cell biology, 188(5), 681-692 (2010-03-10)
We have analyzed the mitotic function of SENP6, a small ubiquitin-like modifier (SUMO) protease that disassembles conjugated SUMO-2/3 chains. Cells lacking SENP6 showed defects in spindle assembly and metaphase chromosome congression. Analysis of kinetochore composition in these cells revealed that
Neil Hattersley et al.
Molecular biology of the cell, 22(1), 78-90 (2010-12-15)
Promyelocytic leukemia protein (PML) is the core component of PML-nuclear bodies (PML NBs). The small ubiquitin-like modifier (SUMO) system (and, in particular, SUMOylation of PML) is a critical component in the formation and regulation of PML NBs. SUMO protease SENP6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service