Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93750

Sigma-Aldrich

(+)-Tubocurarine chloride pentahydrate

≥97.0% (TLC)

Synonym(s):

(+)-Tubocurarine chloride hydrochloride pentahydrate, D-Tubocurarine dichloride pentahydrate, Tubarine pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H42Cl2N2O6 · 5H2O
CAS Number:
Molecular Weight:
771.72
Beilstein:
3896374
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

plant

Quality Level

Assay

≥97.0% (TLC)

form

powder

optical activity

[α]20/D +195±5°, c = 0.5% in H2O

impurities

~12% water

mp

275-280 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].Cl[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].COc1cc2CCN(C)[C@H]3Cc4ccc(Oc5c(O)c(OC)cc6CC[N+](C)(C)[C@H](Cc7ccc(O)c(Oc1cc23)c7)c56)cc4

InChI

1S/C37H40N2O6.2ClH.5H2O/c1-38-14-12-24-19-32(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-37-35-25(20-34(43-5)36(37)41)13-15-39(2,3)29(35)17-23-8-11-30(40)31(18-23)45-33;;;;;;;/h6-11,18-21,28-29H,12-17H2,1-5H3,(H-,40,41);2*1H;5*1H2/t28-,29+;;;;;;;/m0......./s1

InChI key

WMIZITXEJNQAQK-GGDSLZADSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(+)-Tubocurarine chloride pentahydrate has been used:
  • as an acetylcholine receptor antagonist in synapse blocking experiment to study its effect on neuromuscular junction formation (NMJ) formation in a co-culture system of human skeletal muscles and human stem cell-derived motoneurons
  • to induce paralysis in zebrafish larvae to study whole-brain imaging of seizures by two-photon light-sheet microscopy
  • to block NMJ to study its role in myotube contraction

Biochem/physiol Actions

(+)-Tubocurarine chloride pentahydrate serves as a neuromuscular blocker.
Classical competitive antagonist of the nicotinic acetylcholine receptor. Structure-activity relationships and conformational studies for tubocurarine binding at the nicotinic AChR.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Karpen et al.
Biochemistry, 25(7), 1786-1792 (1986-04-08)
The issue of whether d-tubocurarine, the classical acetylcholine receptor inhibitor, inhibits the receptor by a competitive or noncompetitive mechanism has long been controversial. d-Tubocurarine, in this study, has been found to be both a competitive (KC = 120 nM) and
Fast whole-brain imaging of seizures in zebrafish larvae by two-photon light-sheet microscopy
de Vito G, et al.
Biomedical Optics Express, 1516-1536 null
Richard Mills et al.
Molecular metabolism, 7, 12-22 (2017-11-22)
We examined whether skeletal muscle overexpression of PGC-1α1 or PGC-1α4 affected myokine secretion and neuromuscular junction (NMJ) formation. A microfluidic device was used to model endocrine signaling and NMJ formation between primary mouse myoblast-derived myotubes and embryonic stem cell-derived motor
Xiufang Guo et al.
Biomaterials, 32(36), 9602-9611 (2011-09-29)
Functional in vitro models composed of human cells will constitute an important platform in the next generation of system biology and drug discovery. This study reports a novel human-based in vitro Neuromuscular Junction (NMJ) system developed in a defined serum-free
Pietro Ricci et al.
Journal of biophotonics, 15(4), e202100256-e202100256 (2022-01-10)
Two-photon (2P) excitation is a cornerstone approach widely employed in neuroscience microscopy for deep optical access and sub-micrometric-resolution light targeting into the brain. However, besides structural and functional imaging, 2P optogenetic stimulations are less routinary, especially in 3D. This is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service