Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50200

Sigma-Aldrich

Gly-Gly

≥99.0% (NT), suitable for ligand binding assays, BioXtra

Synonym(s):

Diglycine, Glycyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH2COOH
CAS Number:
Molecular Weight:
132.12
Beilstein:
1765223
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Product Name

Gly-Gly, BioXtra, ≥99.0% (NT)

product line

BioXtra

Assay

≥99.0% (NT)

form

powder

technique(s)

ligand binding assay: suitable

ign. residue

≤0.1% (as SO4)

loss

≤1% loss on drying, 110 °C

color

white

useful pH range

7.5-8.9

pKa (25 °C)

8.2

mp

220-240 °C (dec.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NCC(=O)NCC(O)=O

InChI

1S/C4H8N2O3/c5-1-3(7)6-2-4(8)9/h1-2,5H2,(H,6,7)(H,8,9)

InChI key

YMAWOPBAYDPSLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glycine is a non-essential amino acid. Influx of calcium through the cell membrane is mediated by glycine-gated channel. Glycine participates in the synthesis of porphyrins, purine and serine. It also serves as a competitive agonist for glutamate in binding to the NMDA (N-methyl-D-aspartate) receptors. Glycine synthesis might be increased in rapidly proliferating cancer cells, due to increased demand for the amino acid. Diglycine is known to catalyze the formation of homo- and hetero dipeptides more efficiently than glycine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycine and diglycine as possible catalytic factors in the prebiotic evolution of peptides.
Plankensteiner K
Origins of Life and Evolution of the Biosphere : the Journal of the International Society For the Study of the Origin of Life, 32(3), 225-236 (2002)
Metabolite Profiling Identifies a Key Role for Glycine in Rapid Cancer Cell Proliferation
Mohit Jain
Science, 336(6084), 1040-1044 (2012)
Namrata D Udeshi et al.
Molecular & cellular proteomics : MCP, 12(3), 825-831 (2012-12-26)
Detection of endogenous ubiquitination sites by mass spectrometry has dramatically improved with the commercialization of anti-di-glycine remnant (K-ε-GG) antibodies. Here, we describe a number of improvements to the K-ε-GG enrichment workflow, including optimized antibody and peptide input requirements, antibody cross-linking
P B Armentrout et al.
Journal of the American Society for Mass Spectrometry, 23(4), 632-643 (2011-09-29)
We present a full molecular description of fragmentation reactions of protonated diglycine (H(+)GG) by studying their collision-induced dissociation (CID) with Xe using a guided ion beam tandem mass spectrometer (GIBMS). Analysis of the kinetic energy-dependent CID cross sections provides the
P B Armentrout et al.
Journal of the American Society for Mass Spectrometry, 23(4), 621-631 (2011-09-29)
We present a full computational description of the fragmentation reactions of protonated diglycine (H(+)GG). Relaxed potential energy surface scans performed at B3LYP/6-31 G(d) or B3LYP/6-311 + G(d,p) levels are used to map the reaction coordinate surfaces and identify the transition states (TSs) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service