Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48417

Supelco

Acenaphthene-d10 solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D10
CAS Number:
Molecular Weight:
164.27
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

C1(C(c2c3c(c(c(c2[2H])[2H])[2H])c(c(c(c31)[2H])[2H])[2H])([2H])[2H])([2H])[2H]

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2/i1D,2D,3D,4D,5D,6D,7D2,8D2

InChI key

CWRYPZZKDGJXCA-WHUVPORUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acenaphthene-d10 is a deuterated polycyclic aromatic hydrocarbon (PAH) that belongs to ubiquitous class of anthropogenic organic pollutants.

Application

Acenaphthene-d10 may be used as one of the internal standard for the determination of pesticides in cabbage and radish using QuEChERS sample preparation and gas chromatography mass spectrometry method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A multiresidue method for the determination of 107 pesticides in cabbage and radish using QuEChERS sample preparation method and gas chromatography mass spectrometry.
Nguyen T D, et al.
Food Chemistry, 110(1), 207-213 (2008)
Polycyclic aromatic hydrocarbons (PAHs) in urban surface dust of Guangzhou, China: status, sources and human health risk assessment.
Wang W, et al.
The Science of the Total Environment, 409(21), 4519-4527 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service