Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
biological source
non-animal source
product line
EX-CELL®
form
dry powder
technique(s)
cell culture | mammalian: suitable (suspension)
compatibility
for use with CHO, and CHOZN® GS suspension cell lines
components
L-glutamine: no
shipped in
ambient
storage temp.
2-8°C
General description
Application
- EX-CELL®Advanced HD Perfusion Medium has been specifically developed to support growth and productivity across a diverse set of Chinese Hamster Ovary (CHO) cells in perfusion cultures while still allowing for flexibility in the adjustment of protein quality attributes..
- This medium was developed using a strategy of Multivariate Analysis to establish correlations in which the formulation components influence critical processes and product attributes such as titer, fold expansion, protein expression and glycosylation.
- While the medium was developed specifically for steady-state perfusion, it can also support other perfusion applications such as N-1, continuous fed-batch and production perfusion where cells are not bled.
Preparation Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description powder | description CHO Fed-batch Medium. | description CHO Feed 1 (with glucose). Use 64.10 g/L. See information sheet, 24367C @ www.sigmaaldrich.com for hydration instructions | description CHO Feed 1 (without glucose). Use 34.10 g/L. See information sheet, 24368C @ www.sigmaaldrich.com for hydration instructions |
| technique(s) cell culture | mammalian: suitable (suspension) | technique(s) cell culture | mammalian: suitable (suspension) | technique(s) cell culture | mammalian: suitable (suspension) | technique(s) cell culture | mammalian: suitable (suspension) |
| form dry powder | form dry powder | form dry powder | form dry powder |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
| product line EX-CELL® | product line EX-CELL® | product line EX-CELL® | product line EX-CELL® |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| Quality Level 500 | Quality Level 500 | Quality Level 500 | Quality Level 500 |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
WGK
WGK 3
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Difficulty Finding Your Product Or Lot/Batch Number?
How to Find the Product Number
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.




