Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24367C

SAFC

EX-CELL® Advanced CHO Feed 1 (with glucose)

Pharma Manufacturing

Synonym(s):

EX-CELL® Advanced CHO Feed 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

product line

EX-CELL®

Quality Level

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EX-CELL® Advanced CHO Feed 1 (with glucose) is chemically-defined and contains no animal derived components. This feed is intended to be a part of a fed-batch platform, and used with EX-CELL® Advanced CHO Fed-batch Medium (14366C or 24366C). This newest platform from SAFC has been specifically developed to support growth and productivity across a diverse set of Chinese Hamster Ovary (CHO) cells in fed-batch cultures while still allowing for flexibility in the adjustment of protein quality attributes. This contemporary fed-batch platform was developed using a strategy of Multivariate Analysis to establish correlations in which the formulation components influence critical process and product attributes such as titer, fold expansion, receptor expression, and glycosylation. This platform contains no hypoxanthine and thymidine, L-glutamine, phenol red or 2-mercaptoethanol (2-ME).

Linkage

Submit the EX-CELL Advanced CHO Fed-batch request form to get in touch with a SAFC representative.

Quantity

Formulated to contain 64.1 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In this study, nine commercially available, chemically defined media and feed combinations from different vendors were assessed for producing monoclonal antibody in Chinese hamster ovary (CHO) cells.

Why optimize your upstream bioprocess media and feed? This article discusses the why and how of optimizing your media and feed to maximize cell viability and productivity.

This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.

This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.

See All

Related Content

Eliminate the need to clean, autoclave and assemble your traditional glass bioreactors and save days of downtime with the Mobius® 3L Single-use Bioreactor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service