Skip to Content
MilliporeSigma

Skip To

24366C

EX-CELL® Advanced CHO Fed-batch Medium

powder

Synonym(s):

EX-CELL® Advanced CHO Fed-batch Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.71
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

EX-CELL® Advanced, CHO Fed-batch Medium. 20.09 g/L. See information sheet, 24366C @ www.sigmaaldrich.com for hydration instructions.

Quality Segment

product line

EX-CELL®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

shipped in

ambient

storage temp.

2-8°C

General description

EX-CELL® AdvancedCHO Fed-batch Medium is chemically-defined and contains no animal derived components. This platform contains no hypoxanthine andthymidine, L-glutamine, phenol red or 2-mercaptoethanol (2-ME). This medium isintended to be a part of a fed-batch platform and used with EX-CELL® AdvancedCHO Feed 1 (24367C - with glucose or 24368C - without glucose).

Application

  • This newest platform from SAFC has been specifically developed to support growth and productivity across a diverse set of Chinese Hamster Ovary (CHO) cells in fed-batch cultures while still allowing for flexibility in the adjustment of protein quality attributes.
  • This contemporary fed-batch platform was developed using a strategy of Multivariate Analysis to establish correlations in which the formulation components influence critical process and product attributes such as titer, fold expansion, receptor expression, and glycosylation.

Preparation Note

Formulated to contain 22.09 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14366C24360C24370C
technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

form

dry powder

form

liquid

form

dry powder

form

dry powder

product line

EX-CELL®

product line

EX-CELL®

product line

EX-CELL®

product line

EX-CELL®

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500


Still not finding the right product?

Explore all of our products under EX-CELL® Advanced


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters