Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11012010

ESO51 cell line human

Synonym(s):

ESO51 Human Cells, Human ESO51 Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human esophagus

Quality Level

growth mode

Suspension

karyotype

Not specified

morphology

Large spherical clumps of cells growing in suspension

products

P53 mutation c.524G>A

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Distal oesophageal adenocarcinoma,

Application

For complete product information, please see ECACC
Recommended culture medium: RPMI-1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

DNA Profile

STR-PCR Data: Amelogenin: X,
CSF1PO: 10
D13S317: 11
D16S539: 13
D5S818: 11
D7S820: 8.2
THO1: 9
TPOX: 8
vWA: 14,15

Culture Medium

RPMI-1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-9 x 105 cells/cm3 by addition of an equal volume of fresh medium every other day. The DMSO must be removed by centrifugation of the cells at resuscitation. 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The biological meaning of intestinal metaplasia of the gastroesophageal junction.
Paula Chaves et al.
International journal of surgical pathology, 18(3 Suppl), 43S-47S (2010-05-28)
Jurjen J Boonstra et al.
Journal of the National Cancer Institute, 102(4), 271-274 (2010-01-16)
For decades, hundreds of different human tumor type-specific cell lines have been used in experimental cancer research as models for their respective tumors. The veracity of experimental results for a specific tumor type relies on the correct derivation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service