Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1301 Cell Line human

1051619, human blood, Lymphoblast

Synonym(s):

1301 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

1301 Cell Line human, 01051619, T-cell leukemia

biological source

human blood

growth mode

Suspension

karyotype

Tetraploid

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T-cell leukaemia

Cell Line Description

1301 is a sub-line of CCRF-CEM, Sigma catalogue no. 85112105. It has unusually long telomeres, useful in tests where telomere length is determined, where it can be used as control cells with known telomere length. This cell line has been recommended as a control cell line for use with the Telomere PNA Kit/FITC for flow cytometry from DakoCytomation.

Application

Internal control cell in telomere measurements performed by flow cytometry.

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 37°C; 5% CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service