Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

C3881

Sigma-Aldrich

Calcium chloride dihydrate

ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder

pH

5-8 (25 °C, 147 g/L)

mp

176 °C (dec.) (lit.)

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Calcium chloride dihydrate is an inorganic compound that is commonly used as an environmentally friendly catalyst in organic synthesis. It is also used in asymmetric transformation reactions.

Application

Calcium chloride dihydrate can be used as a catalyst:      
  • In the asymmetric 1,4-addition and [3+2]-cycloaddition reactions in the presence of box-type ligands.      
  • To synthesize hexahydropyrimidine derivatives via a one-pot three-component condensation reaction of ethyl/methyl acetoacetate, aniline, and formaldehyde.

It can also be used as a calcium precursor to synthesize macro/mesoporous dicalcium phosphate monolith (CaHPO4) via the sol-gel method using phosphoric acid (H3PO4), and poly(acrylic acid) (PAA).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dante Pascual Goy et al.
The open biomedical engineering journal, 6, 85-91 (2012-08-01)
At present, typical approaches employed to repair fractures and other bone lesions tend to use matrix grafts to promote tissue regeneration. These grafts act as templates, which promote cellular adhesion, growth and proliferation, osteoconduction, and even osteoinduction, which commonly results
Sophie Lanone et al.
Particle and fibre toxicology, 6, 14-14 (2009-05-02)
A critical issue with nanomaterials is the clear understanding of their potential toxicity. We evaluated the toxic effect of 24 nanoparticles of similar equivalent spherical diameter and various elemental compositions on 2 human pulmonary cell lines: A549 and THP-1. A
Danielle Wu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(29), 12096-12101 (2013-07-03)
Osteocytes are bone cells that form cellular networks that sense mechanical loads distributed throughout the bone tissue. Interstitial fluid flow in the lacunar canalicular system produces focal strains at localized attachment sites around the osteocyte cell process. These regions of
Chao-Lin Chen et al.
Scientific reports, 9(1), 10616-10616 (2019-07-25)
Autologous vascular grafts have the advantages of better biocompatibility and prognosis. However, previous studies that implanted bare polymer tubes in animals to grow autologous tubular tissues were limited by their poor yield rates and stability. To enhance the yield rate
Kevin T Urak et al.
Methods (San Diego, Calif.), 103, 167-174 (2016-03-15)
Aptamers are single-stranded DNA or RNA oligonucleotides that can bind with exquisitely high affinity and specificity to target molecules and are thus often referred to as 'nucleic acid' antibodies. Oligonucleotide aptamers are derived through a process of directed chemical evolution

Protocols

Follow our procedure for the determination of a chymotrypsin activity. This enzymatic assay of alpha chymotrypsin guides you through the entire process and necessary calculations.

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service