Skip to Content
MilliporeSigma
All Photos(3)

Documents

M0250

Sigma-Aldrich

Magnesium chloride hexahydrate

ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

solid

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Magnesium chloride hexahydrate is a hydrated form of magnesium chloride. It is used as a catalyst in organic synthesis. In molecular biology, it is used as a source of magnesium ions.

Application

Magnesium chloride hexahydrate can be used as:      
  • A catalyst to synthesize dihydropyrimidine derivatives by Biginelli condensation of aldehydes, 1,3?dicarbonyl compounds, and urea.      
  • A precursor salt to prepare Mg-Fe bi-metal oxide catalyst by treating with iron(II) sulfate heptahydrate. Mg-Fe bi-metal oxide is applicable as a renewable solid base catalyst to synthesize 2-aminothiophenes via Gewald′s reaction.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Günther Gratzl et al.
Colloids and surfaces. B, Biointerfaces, 126, 98-105 (2014-12-30)
The availability of polymeric antimicrobially active surfaces, which are mainly based on cationic surface effects, is limited. We have previously reported the discovery that, in addition to cationic surfaces, anionic surfaces based on poly(acrylic acid) (PAA) copolymers have a bactericidal
Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve
Satyabrata Das et al.
Nature biotechnology, 38(3), 297-302 (2020-02-26)
The scarcity of donor organs may be addressed in the future by using pigs to grow humanized organs with lower potential for immunological rejection after transplantation in humans. Previous studies have demonstrated that interspecies complementation of rodent blastocysts lacking a
Monica Mazzarino et al.
Analytica chimica acta, 837, 70-82 (2014-07-09)
This article describes a liquid chromatographic/tandem mass spectrometric method, based on the use of precursor ion scan as the acquisition mode, specifically developed to detect indole-derived cannabinoids (phenylacetylindoles, naphthoylindoles and benzoylindoles) in biological fluids (saliva, urine and blood). The method
Ikuo Yamamiya et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1485-1492 (2014-07-09)
Tegafur (FT), a prodrug of 5-fluorouracil, is a chiral molecule, a racemate of R- and S-isomers, and CYP2A6 plays an important role in the enantioselective metabolism of FT in human liver microsomes (R-FT > S-FT). This study examined the enantioselective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service