Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

M2670

Sigma-Aldrich

Magnesium chloride hexahydrate

BioXtra, ≥99.0%

Synonym(s):

MgCl2 hexahydrate, MgCl2.6H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
solubility:
H2O: 1 M, clear, colorless

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0%

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

solubility

H2O: 1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ba: ≤0.005%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mn: ≤0.0005%
NH4+: ≤0.002%
Na: ≤0.005%
Pb: ≤0.001%
Sr: ≤0.005%
Zn: ≤0.0005%

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium chloride hexahydrate has been used:
  • in the extraction buffer preparation for determining the enzymatic activity of ADP-glucose pyrophosphorylase (AGPase) and α-amylase
  • in neurobasal culture medium for viability test and in phosphate buffer saline (PBS) for electrophysiological test
  • in the preparation of brine solution
  • in the preparation of the adenosine triphosphate (ATP) –MgCl2 (magnesium chloride)solution

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron is required for zebrafish embryogenesis
Rowe R I and Eckhert C D
The Journal of Experimental Biology, 202(12), 1649-1654 (1999)
Persistent digital hyperthermia over a 48 h period does not induce laminitis in horses
de L M A, et al.
The Veterinary Journal, 192(3), 435-440 (2012)
Microelectrode array-evaluation of neurotoxic effects of magnesium as an implantable biomaterial
Huang T, et al.
Journal of materials science & technology, 32(1), 89-96 (2016)
Influence of tryptophan and indole-3-acetic acid on starch accumulation in the synthetic mutualistic Chlorella sorokiniana-Azospirillum brasilense system under heterotrophic conditions
Palacios O A, et al.
Research in Microbiology, 167(5), 367-379 (2016)
Peter H L Krijger et al.
Methods (San Diego, Calif.), 170, 17-32 (2019-07-29)
Chromosome conformation capture (3C) methods measure DNA contact frequencies based on nuclear proximity ligation, to uncover in vivo genomic folding patterns. 4C-seq is a derivative 3C method, designed to search the genome for sequences contacting a selected genomic site of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service