Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87368

Sigma-Aldrich

Tetrahydrofuran

≥99.9%, ACS reagent, contains ~0.025% 2, 6-di-tert-butyl-4-methylphenol as stabilizer, 250 ppm BHT as inhibitor, suitable for solid phase extraction (SPE), reag. Ph. Eur.

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Assay:
≥99.9%
technique(s):
solid phase extraction (SPE): suitable
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Product Name

Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as inhibitor
~0.025% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

1.8-11.8 %

technique(s)

solid phase extraction (SPE): suitable

impurities

≤0.001% free acid (as CH3COOH)
≤0.005% peroxides (as H2O2)
≤0.05% water

evapn. residue

≤0.001%

refractive index

n20/D 1.407 (lit.)
n20/D 1.407

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.887 g/mL at 20 °C
0.889 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is tetrahydrofuran (THF) and it contains 250ppm BHT (Butylated hydroxytoluene) as an inhibitor. THF is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THF forms double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1,4-diol is formed as an intermediate during the synthesis of THF. Hot THF is useful for the dissolution of polyvinylidene chloride (PVDV).

Application

Tetrahydrofuran has been used as a solvent for the preparation of sol-gel solution for coating solid phase microextraction (SPME) fibers in a study.

It may be used for the dissolution of poly-ε-caprolactone (PCL) and 1,3-diaminopentane, during the preparation of poly-ε-caprolactone (PCL)-hydroxyapatite (HA) scaffolds and acrylate-terminated poly(5-amino-1-pentanol-co-1,4-butanediol diacrylate) (C32)- 1,3-diaminopentane (117) polymer, respectively.

Analysis Note

miscible with water and ethanol (96%) pass

Other Notes

The article number 87368-6X1L-M will be discontinued. Please order the single bottle 87368-1L-M which is physically identical with the same exact specifications.
Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrospun amino-functionalized PDMS as a novel SPME sorbent for the speciation of inorganic and organometallic arsenic species.
Boyaci E, et al.
Royal Society of Chemistry Advances, 3(44), 22261-22268 (2013)
Todd J Harris et al.
Biomaterials, 31(5), 998-1006 (2009-10-24)
The use of biomaterials for gene delivery can potentially avoid many of the safety concerns with viral gene delivery. However, the efficacy of polymeric gene delivery methods is low, particularly in vivo. One significant concern is that the interior and
Haiying Yu et al.
Biomaterials, 30(4), 508-517 (2008-11-01)
Natural bone growth greatly depends on the precedent vascular network that supplies oxygen and essential nutrients and removes metabolites. Likewise, it is crucial for tissue-engineered bone to establish a vascular network that temporally precedes new bone formation, and spatially originates
Polyhedral clathrate hydrates. X. Structure of the double hydrate of tetrahydrofuran and hydrogen sulfide.
Mak TCW, et al.
J. Chem. Phys. , 42(8), 2732-2737 (1965)
Synthetic routes to tetrahydrofuran, tetrahydropyran, and spiroketal units of polyether antibiotics and a survey of spiroketals of other natural products.
Boivin TLB.
Tetrahedron, 43(15), 3309-3362 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service