Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71500

Sigma-Aldrich

Sodium phosphate monobasic dihydrate

purum p.a., crystallized, ≥99.0% (T)

Synonym(s):

Sodium dihydrogen phosphate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

purum p.a.

Quality Level

Assay

≥99.0% (T)

form

crystals

quality

crystallized

pH

4.0-4.5 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1

InChI key

VBJGJHBYWREJQD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dihydrogen phosphate is an inorganic compound, which is soluble in water. Sodium dihydrogen phosphate dihydrate is used as a reagent in the development of silicate-based grouts.

Application

Sodium phosphate monobasic dihydrate has been used:
  • to perform perfusion for fixed brain tissue to the validate magnetic resonance (MR) signal changes and analyze neuropathological changes in the brain
  • as a component of basal salt solution to prepare biofilm minimal media (BMM) for culturing Pseudomonas aeruginosa strain PAO1
  • to prepare sodium phosphate monobasic (NaH2PO4) stock solutions to make running buffer for capillary electrophoresis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Escuder-Gilabert et al.
Journal of chromatography. A, 1363, 331-337 (2014-07-22)
The design of experiments (DOE) is a good option for rationally limiting the number of experiments required to achieve the enantioresolution (Rs) of a chiral compound in capillary electrophoresis. In some cases, the modeled Rs after DOE analysis can be
Wenhu Zhou et al.
Analytical chemistry, 87(7), 4001-4007 (2015-03-11)
RNA-cleaving DNAzymes provide a unique platform for developing biosensors. However, a majority of the work has been performed in clean buffer solutions, while the activity of some important DNAzymes in biological sample matrices is still under debate. Two RNA-cleaving DNAzymes
Vivien Csikós et al.
Archives of toxicology, 94(9), 3297-3313 (2020-05-31)
Deoxynivalenol (DON) or vomitoxin, is a trichothecene mycotoxin produced mainly by Fusarium graminearum and culmorum. Mycotoxins or secondary metabolic products of mold fungi are micro-pollutants, which may affect human and animal health. The neuronal and behavioural actions of DON were
Jan Muendges et al.
Biotechnology progress, 31(4), 925-936 (2015-04-11)
This article presents results of continuous multistage aqueous two-phase extraction of an immunoglobulin G1 from cell supernatant in a mixer-settler unit. An aqueous two-phase system consisting of polyethylene glycol 2000, phosphate salt, and water was applied without and with sodium
Ceren Erdem et al.
Artificial cells, nanomedicine, and biotechnology, 42(4), 237-244 (2013-06-26)
In the present work, we designed an amperometric glucose biosensor based on nickel oxide nanoparticles (NiONPs)-modified carbon paste electrode. The biosensor was prepared by incorporation of glucose oxidase and NiONPs into a carbon paste matrix. It showed good analytical performances

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service