Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439215

Sigma-Aldrich

Tetrahydrofuran

≥99.9%, suitable for HPLC, inhibitor-free

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
Assay:
≥99.9%
technique(s):
HPLC: suitable
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Product Name

Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable

impurities

≤0.015% peroxide (as H2O2)
<0.02% water

evapn. residue

<0.0005%

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

solubility

H2O: soluble

density

0.889 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 212 nm Amax: 1.0
λ: 250 nm Amax: 0.180
λ: 300 nm Amax: 0.020
λ: 350-400 nm Amax: 0.005

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is a saturated cyclic ether with a potential use as a biofuel. Its combustion studies have been reported. Reports suggest it is a better promoter than 1,3 dioxolane for CO2-hydrate formation. The effect of THF on the biological systems has been studied.

Application

Tetrahydrofuran may be used as a solvent in the following processes:
  • Formation of diacetylinic polymers.
  • RAFT polymerization of p-acetoxystyrene.
  • Synthesis of di-tert-butyl-Phosphinoferrocene.
It may be used in the following processes:
  • As mobile phase solvent in high-performance liquid chromatography.
  • As a solvent in the preparation of spin-coated poly(bisphenol A decane ether).
  • Formation of butyrolactone (BTL) by green oxidation method.
  • As a solvent for lignin depolymerization to isolate phenolic monomer.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

recommended

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of tetrahydrofuran to butyrolactone catalyzed by iron-containing clay.
Ausavasukhi A and Sooknoi T.
Green Chemistry, 17(1), 435-441 (2015)
Novel Diacetylinic Aryloxysilane Polymers: A New Thermally Cross-Linkable High Temperature Polymer System.
Drake K, et al.
Macromolecules, 46(11), 4370-4377 (2013)
1,3 Dioxolane versus tetrahydrofuran as promoters for CO2-hydrate formation: Thermodynamics properties, and kinetics in presence of sodium dodecyl sulfate.
Torre JP, et al.
Chemical Engineering Science, 126, 688-697 (2015)
Practical Synthesis of Di-tert-Butyl-Phosphinoferrocene.
Busacca CA, et al.
Organic Syntheses, 90, 316-326 (2013)
Solène I Cauët et al.
Journal of polymer science. Part A, Polymer chemistry, 48(12), 2517-2524 (2010-07-27)
The kinetics of the RAFT polymerization of p-acetoxystyrene using a trithiocarbonate chain transfer agent, S-1-dodecyl-S'-(α,α'-dimethyl-α″-acetic acid)trithiocarbonate, DDMAT, was investigated. Parameters including temperature, percentage initiator, concentration, monomer-to-chain transfer agent ratio and solvent were varied and their impact on the rate of

Articles

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service