Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31630

Sigma-Aldrich

Silver nitrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

5.8 (vs air)

Assay

≥99.8%

form

solid

impurities

≤0.01% subst. not precipitated by HCl (as SO4)

mp

212 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrite (NO2-): ≤500 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Cd: ≤1 mg/kg
Cu: ≤2 mg/kg
Fe: ≤2 mg/kg
Mn: ≤5 mg/kg
Pb: ≤10 mg/kg
Zn: ≤1 mg/kg

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lee-Meisel method has been proposed for the preparation of silver colloids for surface-enhanced Raman scattering (SERS), via AgNO3 reduction with sodium citrate. It has been reported as an antagonist of ethylene.

Application

Silver nitrate may be employed as stain to study the osteogenic differentiation of fetal human mesenchymal stem/progenitor cells (MSCs). It may be used in the preparation of highly sensitive silver colloids for surface-enhanced Raman scattering (SERS) studies.

Disclaimer

The product is notintended for use as a biocide under global biocide regulations, including butnot limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act,European Biocidal ProductsRegulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal ProductsRegulation, Korea’s Consumer Chemical Products and BiocideSafety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D D Songstad et al.
Plant cell reports, 7(4), 262-265 (1988-06-01)
The effect of the ethylene antagonists norbornadiene and silver nitrate and the ethylene precursor l-aminocyclopropane-l-carboxylic acid (ACC) on Zea mays plant regeneration was studied. A 12-fold increase in plant regeneration, as measured by number of plants obtained per gram fresh
A new method for fast preparation of highly surface-enhanced Raman scattering (SERS) active silver colloids at room temperature by reduction of silver nitrate with hydroxylamine hydrochloride.
Leopold N and Lendl B.
The Journal of Physical Chemistry B, 107(24), 5723-5727 (2003)
C Campagnoli et al.
Blood, 98(8), 2396-2402 (2001-10-06)
Human mesenchymal stem/progenitor cells (MSCs) have been identified in adult bone marrow, but little is known about their presence during fetal life. MSCs were isolated and characterized in first-trimester fetal blood, liver, and bone marrow. When 10(6) fetal blood nucleated
Lara Settimio et al.
Environmental pollution (Barking, Essex : 1987), 191, 151-157 (2014-05-20)
The fate and lability of added soluble Ag in soils over time was examined by measurement of labile metal (E-value) by isotopic dilution using the (110m)Ag radioactive isotope and the solid-phase speciation of Ag by X-ray absorption near edge structure
Tao Xu et al.
Organic letters, 14(21), 5416-5419 (2012-10-24)
A silver-catalyzed intramolecular oxidative aminofluorination of alkynes has been developed by using NFSI as a fluorinating reagent. This reaction represents an efficient method for the synthesis of various 4-fluoroisoquinolines and 4-fluoropyrrolo[α]isoquinolines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service