Skip to Content
MilliporeSigma
Silver nitrate tested according to Ph. Eur.

Silver nitrate

Silver nitrate tested according to Ph. Eur.
Synonym(s):
Nitric acid silver(I) salt, Silver nitrate
Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade