MilliporeSigma
Fate and lability of silver in soils: effect of ageing.
Environmental pollution (Barking, Essex : 1987) (2014-05-20)
Lara Settimio, Mike J McLaughlin, Jason K Kirby, Kate A Langdon, Enzo Lombi, Erica Donner, Kirk G Scheckel
ABSTRACT
The fate and lability of added soluble Ag in soils over time was examined by measurement of labile metal (E-value) by isotopic dilution using the (110m)Ag radioactive isotope and the solid-phase speciation of Ag by X-ray absorption near edge structure (XANES) spectroscopy. After two weeks of ageing the E-values for Ag decreased by 20-90% with a further decrease of 10-40% after six months. The overall decrease in labile Ag for all soils after the 6 month ageing period was 50-100%. The ageing was more rapid and pronounced in the alkaline soils. XANES results for Ag in soils indicated that for the majority of soils the added Ag(+) was reduced to metallic Ag over time, and associations with Fe-oxohydroxides and reduced S groups in organic matter also decreased Ag lability. Strong positive correlations were found between metallic Ag and non-labile Ag and between organic carbon and Ag bonded with S species.
Sigma-Aldrich
Silver nitrate solution, 0.0282 M
Sigma-Aldrich
Silver nitrate solution, 0.05 M
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)