Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31606

Sigma-Aldrich

Cerium(IV) sulfate tetrahydrate

puriss. p.a., ≥98%

Synonym(s):

Ceric sulfate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(SO4)2 · 4H2O
CAS Number:
Molecular Weight:
404.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
powder

vapor pressure

<0.01 hPa ( 20 °C)
<0.01 hPa ( 50 °C)

Quality Level

grade

puriss. p.a.

Assay

≥98%

form

powder

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤0.01% insoluble matter in dilute H2SO4

pH

~0.5 (20 °C, 207 g/L)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤100 mg/kg

cation traces

Cu: ≤10 mg/kg
Fe: ≤50 mg/kg
Pb: ≤10 mg/kg

SMILES string

[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Ce.2H2O4S/c;2*1-5(2,3)4/h;2*(H2,1,2,3,4)/q+4;;/p-4

InChI key

VZDYWEUILIUIDF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(IV) sulfate tetrahydrate is a hydrated salt of cerium sulfate. Its molecule contains cerium ions coordinated with eight oxygen atoms. Its crystals have been reported to be orthorhombic.

Application

Cerium(IV) sulfate tetrahydrate may be employed in the synthesis of alkyl esters of oxo acids and oxo acids, via reaction with 2-substituted cycloalkanone. It may be used in the preparation of 0.01M cerium(IV) sulfate in 2M sulfuric acid solution.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyuki Miura et al.
Journal of chromatography. A, 956(1-2), 77-84 (2002-07-11)
Rapid separation and determination of mixtures of L-ascorbic acid, nitrite, sulfite, oxalate, iodide and thiosulfate by conventional ion chromatography is often difficult due to incomplete separation of L-ascorbic acid and nitrite from the water peak when using eluents giving short
The Crystal Structure of Cerium (IV) Sulfate Tetrahydrate.
Lindgren O.
Acta Chemica Scandinavica, Series A: Physical and Inorganic Chemistry, 31(6), 453-456 (1977)
Oxidation of 2-Substituted Cycloalkanones with Cerium (IV) Sulfate Tetrahydrate in Alcohols and Acetic Acid.
He L and Horiuchi CA.
Bulletin of the Chemical Society of Japan, 72(11), 2515-2521 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service