Skip to Content
MilliporeSigma
All Photos(1)

Documents

22269

Sigma-Aldrich

Ammonium cerium(IV) sulfate dihydrate

puriss. p.a., ACS reagent, ≥98.0% (RT)

Synonym(s):

Ceric ammonium sulfate, Cerium(IV) ammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)4(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.03

grade

ACS reagent
puriss. p.a.

Assay

≥98.0% (RT)

form

powder or crystals

reaction suitability

reagent type: catalyst
core: cerium

mp

130 °C (lit.)

anion traces

phosphate (PO43-): ≤300 mg/kg

cation traces

Fe: ≤100 mg/kg

SMILES string

N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Ce.4H3N.4H2O4S.2H2O/c;;;;;4*1-5(2,3)4;;/h;4*1H3;4*(H2,1,2,3,4);2*1H2/q+4;;;;;;;;;;/p-4

InChI key

VCNAMBGKEDPVGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast colorimetric method for measuring urinary iodine.
Daniella Gnat et al.
Clinical chemistry, 49(1), 186-188 (2003-01-01)
T Ohashi et al.
Clinical chemistry, 46(4), 529-536 (2000-04-12)
Urinary iodine is a good biochemical marker for control of iodine deficiency disorders. Our aim was to develop and validate a simple, rapid, and quantitative method based on the Sandell-Kolthoff reaction, incorporating both the reaction and the digestion process into
Thermal behaviour of sulphato and nitrato complexes of cerium (IV).
Pokol G, et al.
J. Therm. Anal., 42(2-3), 343-359 (1994)
S Kaul et al.
Amino acids, 34(2), 315-320 (2006-11-07)
An assessment of the potential of proline to scavenge free radicals was made in a couple of in vitro assay systems, namely graft co-polymerization and autooxidation of pyrogallol. Both these assays are essentially dependent upon free radical mechanisms. Graft co-polymerization
K H Gordon et al.
Organic letters, 3(1), 53-56 (2001-06-30)
[figure: see text] A novel benzyloxyaniline linker that uses ceric ammonium nitrate (CAN) as a cleavage reagent is described. Its application in the solid-phase synthesis of N-unsubstituted beta-lactams and secondary amides furnishes compounds in moderate to excellent yield (45-91%) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service