Skip to Content
MilliporeSigma
All Photos(3)

Documents

258776

Sigma-Aldrich

2,2,4-Trimethylpentane

ReagentPlus®, ≥99%

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.9 (vs air)

Quality Level

vapor pressure

41 mmHg ( 21 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,4-Trimethylpentane (TMP, isooctane) is a saturated, branched hydrocarbon. It is a water-insoluble compound. Isooctane is used in gasoline and aviation fuel to increase the octane level or provide antiknock properties. Additionally, it is used as a solvent, thinner, and in organic synthesis.

Application

2,2,4-Trimethylpentane is used:
  • In the binary mixture (ethane + 2,2,4-trimethylpentane), to analyses the solubility of ethane using static–synthetic apparatus.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quality Prediction from Hydroprocessing through Infrared Spectroscopy (IR).
Orrego-Ruiz JA, et al.
Energy and Fuels, 26(1), 586-593 (2011)
B G Short et al.
Toxicologic pathology, 14(2), 194-203 (1986-01-01)
Unleaded gasoline causes acute and chronic nephrotoxicity and renal tumors in male rats, but not female rats or mice of either sex. An active nephrotoxic component of unleaded gasoline has been identified as 2,2,4-trimethylpentane (TMP). The first objective of this
Physicochemical properties of liquid mixtures. 1. Viscosity, density, surface tension and refractive index of cyclohexane+ 2, 2, 4-trimethylpentane binary liquid systems from 25 C to 50 ?C.
Gomez-Diaz D, et al.
Journal of Chemical and Engineering Data, 46(3), 720-724 (2001)
E A Lock et al.
Toxicology and applied pharmacology, 91(2), 182-192 (1987-11-01)
Trimethylpentane (TMP) produces nephrotoxicity in male but not in female rats. The toxicity is characterized by an increase in protein droplets in proximal convoluted tubular cells and an increase in the renal concentration of the male-rat-specific protein alpha 2u-globulin. Subcellular
Density and viscosity measurements of 2, 2, 4-trimethylpentane (isooctane) from 198 K to 348 K and up to 100 MPa.
Padua AAH, et al.
Journal of Chemical and Engineering Data, 41(6), 1488-1494 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service