Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.21627

Sigma-Aldrich

Isooctane

for synthesis

Synonym(s):

2,2,4-Trimethylpentane, 2,2,4-Trimethylpentane, Isobutyltrimethylmethane, iso-Octane, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
MDL number:
UNSPSC Code:
12352001
EC Index Number:
208-759-1
NACRES:
NA.22
grade:
for synthesis
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

745 °F

potency

>2500 mg/kg LD50, oral (Rat)

expl. lim.

6 %

dilution

(for synthesis)

refractive index

n20/D 1.391 (lit.)

pH

7

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

transition temp

flash point -12 °C

density

0.692 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Green synthesis of beta-sitostanol esters catalyzed by the versatile lipase/sterol esterase from Ophiostoma piceae.: This study explores the use of Isooctane as a solvent in the green synthesis of beta-sitostanol esters. The research demonstrates the effectiveness of isooctane in enhancing the catalytic activity of lipase/sterol esterase, making it a valuable solvent in sustainable chemistry applications for the food industry (Molina-Gutirrez et al., 2017).

  • Single-Step Partial Purification of Intracellular beta-Galactosidase from Kluyveromyces lactis Using Microemulsion Droplets.: This research employs isooctane in the formation of microemulsion droplets for the partial purification of beta-galactosidase. The study highlights the utility of isooctane in biotechnological applications, particularly in enzyme purification and biocatalysis (Mazı et al., 2016).

  • Profiling of urinary amino-carboxylic metabolites by in-situ heptafluorobutyl chloroformate mediated sample preparation and gas chromatography-mass spectrometry.: Isooctane is used as a solvent in the sample preparation process for profiling urinary metabolites. The study showcases isooctane′s role in enhancing the sensitivity and specificity of gas chromatography-mass spectrometry (GC-MS) analyses in clinical and biochemical research (Huek et al., 2016).

  • Extraction of bovine serum albumin using reverse micelles formed by hexadecyl trimethyl ammonium chloride.: Isooctane is utilized in the formation of reverse micelles for the extraction of bovine serum albumin. This research underscores the potential of isooctane in bioprocessing and protein purification, relevant to biochemistry and molecular biology fields (Sun et al., 2011).

Specifications

Saturation Concentration in Air: 239 g/m3 (20 °C)

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.691 - 0.692
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service