Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

199974

Sigma-Aldrich

Aluminum oxide

activated, Brockmann I, standard grade, neutral

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52

form

powder

manufacturer/tradename

CAMAG 507-C-I

technique(s)

ion chromatography: suitable

impurities

<0.03% Fe2O3
<0.03% SiO2
<0.4% Na2O

particle size

40-160 μm

pore size

58 Å pore size

pH

7.0±0.5 ( in H2O)

mp

2040 °C (lit.)

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The mechanism of formation of protective scales of α-Al2O3 on surfaces of binary M-Al (M = Fe, Ni or Co) alloys at high temperatures has been studied. Activated aluminum oxides having different acidities and water concentrations (having Brockmann activity I-V) can easily detect various types of water (namely free water, pore water, and bound water) on the aluminum oxide present in soils.

Application

Aluminum oxide (Alumina, Al2O3) has been used in the preparation of one-step self-assembled double-helices of multiwalled carbon nanotubes (MWCNT) arrays (composed of Fe nanoparticles bound to SiOx micro-particles).
Aluminum oxide (neutral) has been used in the preparation of alumina supported silver (Ag/Al2O3) catalyst suitable for selective catalytic reduction of nitrogen oxides (NOx) by hydrocarbons (HC-SCR).

Packaging

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of Ag O Al entities in adsorption of NCO species and reduction of NO x.
Deng H, et al.
Catalysis Today, 258, 35-40 (2015)
Carbon nanotube bundles self-assembled in double helix microstructures.
Cervantes-Sodi F, et al.
Carbon, 50(10), 3688-3693 (2012)
Prescott R and Graham MJ.
Oxidation of Metals, 38(3-4), 233-254 (1992)
Thermoanalytical studies of water on activated alumina, Brockmann I-V,(acid, neutral, basic) from- 60? to+ 700? C.
Hampson JW and Bleam WF.
Thermochimica Acta, 288(1), 179-189 (1996)
Colin J Ingham et al.
Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service