Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
Al2O3
CAS Number:
Molecular Weight:
101.96
UNSPSC Code:
41115711
EC Index Number:
215-691-6
MDL number:
material
aluminum oxide (irregularly-shaped)
Quality Segment
potency
>5000 mg/kg LD50, oral (Rat)
matrix active group
Irregular-Shaped Aluminum Oxide
particle size
63-200 μm
pore size
9 nm mean pore size
bp
2980 °C/1013 hPa
mp
2050 °C
density
3.94 g/cm3 at 20 °C
application(s)
pharma/biopharma processes
plasma purification
separation technique
normal phase
storage temp.
no temp limit
SMILES string
[Al+3].[Al+3].[O-2].[O-2].[O-2]
InChI
1S/2Al.3O/q2*+3;3*-2
InChI key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
General description
A versatile sorbent for preparative chromatography exhibiting a higher pH stability
Features and Benefits
Aluminum oxide 90 active neutral provides users with:
An aluminum oxide crystal structure comprises octahedrally and tetrahedrally coordinated aluminum coupled by oxygen atoms. The surface is covered by free hydroxyl groups. There are different acidic and basic centers that result in anion and cation exchange properties. Aluminum oxide exhibits a higher pH-stability than silica gel, especially in the alkaline range and occurs in various crystal modifications resulting in different pore diameters.
An aluminum oxide crystal structure comprises octahedrally and tetrahedrally coordinated aluminum coupled by oxygen atoms. The surface is covered by free hydroxyl groups. There are different acidic and basic centers that result in anion and cation exchange properties. Aluminum oxide exhibits a higher pH-stability than silica gel, especially in the alkaline range and occurs in various crystal modifications resulting in different pore diameters.
Packaging
Plastic bottle
Analysis Note
Activity: I
pH-value (10 % suspension): 6.8 - 7.8
Substances extractable by water: ≤ 0.2 %
Chloride (Cl): ≤ 0.1 %
Ca (Calcium): ≤ 0.15 %
Speed of filtration: ≥ 0.3 ml/min
Particle size: 70 % between 0.063 - 0.200 mm
Setting of activity according to the Brockmann activity grades adjusted to a migration distance of 30 mm:
I: activity I
II: activity II
III: activity III
IV: activity IV
V: activity V
pH-value (10 % suspension): 6.8 - 7.8
Substances extractable by water: ≤ 0.2 %
Chloride (Cl): ≤ 0.1 %
Ca (Calcium): ≤ 0.15 %
Speed of filtration: ≥ 0.3 ml/min
Particle size: 70 % between 0.063 - 0.200 mm
Setting of activity according to the Brockmann activity grades adjusted to a migration distance of 30 mm:
I: activity I
II: activity II
III: activity III
IV: activity IV
V: activity V
Storage Class
13 - Non Combustible Solids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.