Skip to Content
MilliporeSigma
All Photos(1)

Documents

13418

Sigma-Aldrich

Sodium carbonate

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Assay

99.5-100.5% (calc. to the dried substance)

form

powder or crystals

quality

meets analytical specification of Ph. Eur., BP, NF, FCC, E500

impurities

alkali hydroxides and bicarbonates, complies
residual solvents
<5 ppm heavy metals (as Pb)

loss

≤0.5% loss on drying, 300 °C
<0.5% loss on drying, 105°C, 4hrs

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤20 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤4 mg/kg
Pb: >2 ppm
Zn: ≤20 mg/kg

suitability

complies for appearance of solution

storage temp.

15-25°C

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Development of novel portable NIR spectroscopy process analytical technology (PAT) tool for monitoring the transition of ibuprofen to ibuprofen sodium during wet granulation process: This study emphasizes the use of sodium carbonate in enhancing the sensitivity and efficiency of NIR spectroscopy tools in pharmaceutical manufacturing, showcasing its pivotal role in real-time quality control (Atanaskova E, Geskovski N, 2024).

  • Physicochemical and antioxidant properties of pectin fractions extracted from lemon (Citrus Eureka) peels: Demonstrates the application of sodium carbonate in the extraction processes of biologically active compounds from citrus peels, highlighting its importance in the development of natural antioxidants for health-related products (Su Z, Li G, 2024).

  • Bryophyte-Bioinspired Nanoporous AAO/C/MgO Composite for Enhanced CO2 Capture: The Role of MgO: This research incorporates sodium carbonate to facilitate the creation of innovative materials designed for efficient CO2 capture, addressing environmental concerns through advanced material science (Cortés-Valadez PJ, Cigarroa-Mayorga OE, 2024).

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Palomero-Gallagher et al.
The Journal of comparative neurology, 508(6), 906-926 (2008-04-12)
Anterior cingulate cortex (ACC) is involved in emotion, mood, and autonomic regulation. Although a subgenual part of ACC (sACC) may be vulnerable in depression and area 25 is cytologically unique, there are no assessments that contrast this region with pregenual
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by
Sandra Nilsson et al.
PloS one, 7(11), e46980-e46980 (2012-11-28)
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of
Yukiyo Sato et al.
PloS one, 9(6), e99041-e99041 (2014-06-11)
The accumulation of RCY1 protein, which is encoded by RESISTANCE TO CMV(Y) (RCY1), a CC-NB-LRR class R-gene, is tightly correlated with the strength of the resistance to a yellow strain of Cucumber mosaic virus [CMV(Y)] in Arabidopsis thaliana. In order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service